Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZC3H14
Entrez Gene ID
79882
Gene Name
zinc finger CCCH-type containing 14
Gene Aliases
MSUT-2, NY-REN-37, SUT2, UKp68
Chromosome Location
Chr.14: 88562909 - 88614720 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001160103.1 NP_001153575.1 3 - 4 417 78
NM_001160104.1 NP_001153576.1 3 - 4 417 78
NM_001326295.1 NP_001313224.1 3 - 4 417 78
NM_001326296.1 NP_001313225.1 3 - 4 417 78
NM_001326297.1 NP_001313226.1 3 - 4 598 78
NM_001326298.1 NP_001313227.1 3 - 4 417 78
NM_001326299.1 NP_001313228.1 3 - 4 417 78
NM_001326301.1 NP_001313230.1 3 - 4 598 78
NM_001326303.1 NP_001313232.1 3 - 4 598 78
NM_001326306.1 NP_001313235.1 3 - 4 417 78
NM_001326307.1 NP_001313236.1 3 - 4 417 78
NM_001326310.1 NP_001313239.1 3 - 4 417 78
NM_001326312.1 NP_001313241.1 3 - 4 417 78
NM_001326313.1 NP_001313242.1 3 - 4 417 78
NM_001326315.1 NP_001313244.1 3 - 4 598 78
NM_001326316.1 NP_001313245.1 3 - 4 265 78
NM_024824.4 NP_079100.2 3 - 4 417 78
NM_207660.3 NP_997543.1 3 - 4 417 78
NM_207661.2 NP_997544.1 3 - 4 265 78
NR_136936.1 3 - 4 417 78
XM_005268067.4 XP_005268124.1 3 - 4 412 78
XM_005268068.4 XP_005268125.1 3 - 4 412 78
XM_005268069.4 XP_005268126.1 3 - 4 412 78
XM_005268070.4 XP_005268127.1 3 - 4 412 78
XM_005268071.4 XP_005268128.1 3 - 4 397 78
XM_011537160.2 XP_011535462.1 3 - 4 525 78
XM_011537161.2 XP_011535463.1 3 - 4 412 78
XM_011537162.2 XP_011535464.1 3 - 4 412 78
XM_017021648.1 XP_016877137.1 3 - 4 412 78
XM_017021649.1 XP_016877138.1 3 - 4 412 78
XM_017021650.1 XP_016877139.1 3 - 4 412 78
GenBank mRNA AK021868.1 - 3 - 4 263 78
AK302136.1 - 3 - 4 343 78
AK303123.1 - 3 - 4 318 78
AK304275.1 - 3 - 4 288 78
AL834215.1 - 3 - 4 265 78
AY578060.1 - 3 - 4 301 78
AY578061.1 - 3 - 4 356 78
AY578062.1 - 3 - 4 265 78
BC011793.1 - 3 - 4 283 78
BX640716.1 - 3 - 4 296 78
DB452844.1 - 3 - 4 417 78

View Full Product Details