The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024827.3 | NP_079103.2 | 3 - 4 | 388 | 111 |
XM_005265491.1 | XP_005265548.1 | 3 - 4 | 353 | 111 | |
XM_006713338.1 | XP_006713401.1 | 3 - 4 | 388 | 111 | |
XM_011534131.1 | XP_011532433.1 | 3 - 4 | 476 | 111 | |
XM_017007234.1 | XP_016862723.1 | 3 - 4 | 476 | 111 | |
GenBank mRNA | AK025890.1 | - | 3 - 4 | 279 | 111 |
AL834223.1 | - | 3 - 4 | 646 | 111 | |
BC009676.1 | - | 2 - 3 | 264 | 111 | |
BG708402.1 | - | 3 - 4 | 358 | 111 | |
DB025431.1 | - | 3 - 4 | 388 | 111 |