The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024843.3 | NP_079119.3 | 1 - 2 | 382 | 56 |
GenBank mRNA | AK027115.1 | - | 1 - 2 | 224 | 56 |
AK225459.1 | - | 1 - 2 | 224 | 56 | |
AK291731.1 | - | 1 - 2 | 232 | 56 | |
AK316588.1 | - | 1 - 2 | 261 | 56 | |
AL136693.1 | - | 1 - 2 | 265 | 56 | |
BC065290.1 | - | 1 - 2 | 217 | 56 | |
CX866825.1 | - | 1 - 2 | 273 | 56 | |
DB235339.1 | - | 1 - 2 | 382 | 56 |