The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024855.3 | NP_079131.3 | 5 - 6 | 1202 | 80 |
GenBank mRNA | AK022847.1 | - | 5 - 6 | 1187 | 80 |
BC001324.1 | - | 4 - 5 | 601 | 80 | |
BC006162.1 | - | 2 - 3 | 384 | 80 | |
BC038402.1 | - | 5 - 6 | 1202 | 80 |