The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024857.4 | NP_079133.3 | 7 - 8 | 3015 | 67 |
XM_011525269.2 | XP_011523571.1 | 7 - 8 | 3036 | 67 | |
XM_011525270.2 | XP_011523572.1 | 7 - 8 | 3036 | 67 | |
XM_011525273.2 | XP_011523575.1 | 7 - 8 | 4053 | 67 | |
XM_011525274.2 | XP_011523576.1 | 2 - 3 | 264 | 67 | |
XM_017025111.1 | XP_016880600.1 | 7 - 8 | 3011 | 67 | |
GenBank mRNA | AJ314648.2 | - | 7 - 8 | 2983 | 67 |
AK022797.1 | - | 6 - 7 | 1510 | 67 | |
AL832103.1 | - | 7 - 8 | 3016 | 67 | |
AY557611.1 | - | 7 - 8 | 2983 | 67 |