Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYO15B
Entrez Gene ID
80022
Gene Name
myosin XVB
Gene Aliases
MYO15BP
Chromosome Location
Chr.17: 75587545 - 75626850 on Build GRCh38
Interrogated Sequence Translated Protein RNADB ID Exon Boundary Assay Location Amplicon Length
RefSeq NM_001309242.1 NP_001296171.1
52 - 53 7972 72
XM_017025120.1 XP_016880609.1
54 - 55 7679 72
XM_017025121.1 XP_016880610.1
54 - 55 7664 72
XM_017025122.1 XP_016880611.1
54 - 55 7643 72
XM_017025123.1 XP_016880612.1
54 - 55 7681 72
XM_017025124.1 XP_016880613.1
53 - 54 7553 72
XM_017025125.1 XP_016880614.1
52 - 53 7402 72
XM_017025126.1 XP_016880615.1
49 - 50 7144 72
XM_017025127.1 XP_016880616.1
52 - 53 6987 72
XM_017025128.1 XP_016880617.1
53 - 54 6733 72
XM_017025129.1 XP_016880618.1
52 - 53 5965 72
XM_017025130.1 XP_016880619.1
51 - 52 5921 72
XM_017025131.1 XP_016880620.1
50 - 51 6360 72
XM_017025132.1 XP_016880621.1
49 - 50 6301 72
XM_017025133.1 XP_016880622.1
47 - 48 5826 72
XM_017025134.1 XP_016880623.1
48 - 49 6187 72
XM_017025135.1 XP_016880624.1
42 - 43 4843 72
XM_017025136.1 XP_016880625.1
42 - 43 5192 72
XM_017025137.1 XP_016880626.1
39 - 40 4737 72
XM_017025138.1 XP_016880627.1
34 - 35 4233 72
XM_017025139.1 XP_016880628.1
35 - 36 4034 72
XM_017025140.1 XP_016880629.1
32 - 33 3847 72
XM_017025143.1 XP_016880632.1
27 - 28 3124 72
XM_017025144.1 XP_016880633.1
48 - 49 7607 72
GenBank mRNA AF418292.1 -
27 - 28 3582 72
AF418293.1 -
28 - 29 3188 72
AK026339.1 -
4 - 5 287 72
AL713730.1 -
3 - 4 181 72
BC027875.1 - LIT3316
MYO15BP
4 - 5 676 72

View Full Product Details