The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308164.1 | NP_001295093.1 | 14 - 15 | 1767 | 72 |
NM_001308166.1 | NP_001295095.1 | 13 - 14 | 1710 | 72 | |
NM_024996.5 | NP_079272.4 | 13 - 14 | 1710 | 72 | |
XM_006713795.1 | XP_006713858.1 | 12 - 13 | 1624 | 72 | |
GenBank mRNA | AF309777.1 | - | 13 - 14 | 1710 | 72 |
AF367998.1 | - | 13 - 14 | 1710 | 72 | |
AK000780.1 | - | 13 - 14 | 1710 | 72 | |
AK092293.1 | - | 12 - 13 | 1316 | 72 | |
AK315031.1 | - | 13 - 14 | 1649 | 72 | |
BC049210.1 | - | 13 - 14 | 1674 | 72 | |
KU178816.1 | - | 13 - 14 | 1603 | 72 | |
KU178817.1 | - | 12 - 13 | 1482 | 72 |