Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 15455792-15456289 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COLQ
Entrez Gene ID
8292
Gene Name
collagen like tail subunit of asymmetric acetylcholinesterase
Gene Aliases
CMS5, EAD
UniGene
Hs.146735
Gene Chromosome Location
Chr.3: 15450133-15521751 on Build GRCh38
Transcripts
NM_005677.3 NM_080538.2 NM_080539.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4114671 c.1135G>A p.G379S Chr.3: 15455929-15455929 on Build GRCh38
5994764 c.1070A>G p.Y357C Chr.3: 15455994-15455994 on Build GRCh38
1593432 c.1143C>T p.S381S Chr.3: 15455921-15455921 on Build GRCh38
6633944 c.1134C>T p.D378D Chr.3: 15455930-15455930 on Build GRCh38
6633942 c.1164C>T p.D388D Chr.3: 15455930-15455930 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769944838 Chr.3: 15456123-15456123 on Build GRCh38
rs754357969 Chr.3: 15456009-15456009 on Build GRCh38
rs970111842 Chr.3: 15455988-15455988 on Build GRCh38
rs1006677510 Chr.3: 15455873-15455873 on Build GRCh38
rs748582765 Chr.3: 15455908-15455908 on Build GRCh38
rs368575417 Chr.3: 15455937-15455937 on Build GRCh38
rs370159858 Chr.3: 15455860-15455860 on Build GRCh38
rs990280097 Chr.3: 15455957-15455957 on Build GRCh38
rs1047967788 Chr.3: 15456059-15456059 on Build GRCh38
rs571916975 Chr.3: 15455854-15455854 on Build GRCh38
rs202019416 Chr.3: 15455977-15455977 on Build GRCh38
rs550132955 Chr.3: 15456054-15456054 on Build GRCh38
rs757951378 Chr.3: 15455875-15455875 on Build GRCh38
rs778085094 Chr.3: 15456008-15456008 on Build GRCh38
rs139329020 Chr.3: 15456256-15456257 on Build GRCh38
rs983479611 Chr.3: 15456163-15456163 on Build GRCh38
rs1043344843 Chr.3: 15456006-15456006 on Build GRCh38
rs779594053 Chr.3: 15455899-15455899 on Build GRCh38
rs1007222915 Chr.3: 15456081-15456081 on Build GRCh38
rs765874335 Chr.3: 15455964-15455964 on Build GRCh38
rs745531202 Chr.3: 15455842-15455842 on Build GRCh38
rs554009695 Chr.3: 15456138-15456139 on Build GRCh38
rs751113924 Chr.3: 15455966-15455966 on Build GRCh38
rs769642026 Chr.3: 15456071-15456071 on Build GRCh38
rs746313049 Chr.3: 15456154-15456154 on Build GRCh38
rs140955551 Chr.3: 15455941-15455941 on Build GRCh38
rs947483883 Chr.3: 15456218-15456218 on Build GRCh38
rs745993482 Chr.3: 15456030-15456030 on Build GRCh38
rs752967536 Chr.3: 15456005-15456005 on Build GRCh38
rs28538922 Chr.3: 15456101-15456101 on Build GRCh38
rs994531614 Chr.3: 15456113-15456113 on Build GRCh38
rs938847804 Chr.3: 15455816-15455816 on Build GRCh38
rs751221973 Chr.3: 15455885-15455885 on Build GRCh38
rs749266800 Chr.3: 15455920-15455920 on Build GRCh38
rs1021787358 Chr.3: 15456229-15456229 on Build GRCh38
rs374437099 Chr.3: 15455864-15455864 on Build GRCh38
rs772677620 Chr.3: 15456158-15456158 on Build GRCh38
rs116373583 Chr.3: 15455986-15455986 on Build GRCh38
rs149852377 Chr.3: 15455930-15455930 on Build GRCh38
rs1031602755 Chr.3: 15456209-15456209 on Build GRCh38
rs938901469 Chr.3: 15456053-15456053 on Build GRCh38
rs951648213 Chr.3: 15456139-15456139 on Build GRCh38
rs74530115 Chr.3: 15456222-15456222 on Build GRCh38
rs765962280 Chr.3: 15455881-15455881 on Build GRCh38
rs192611720 Chr.3: 15456114-15456114 on Build GRCh38
rs1008156537 Chr.3: 15455932-15455932 on Build GRCh38
rs774638847 Chr.3: 15455929-15455929 on Build GRCh38
rs1038178362 Chr.3: 15456112-15456112 on Build GRCh38
rs794727145 Chr.3: 15455918-15455918 on Build GRCh38
rs996172236 Chr.3: 15456145-15456145 on Build GRCh38
rs769982050 Chr.3: 15456012-15456012 on Build GRCh38
rs908011552 Chr.3: 15456096-15456096 on Build GRCh38
rs1008126780 Chr.3: 15456197-15456197 on Build GRCh38
rs755782087 Chr.3: 15455898-15455898 on Build GRCh38
rs996208256 Chr.3: 15455906-15455906 on Build GRCh38
rs765261362 Chr.3: 15455867-15455867 on Build GRCh38
rs561255785 Chr.3: 15456165-15456165 on Build GRCh38
rs890464664 Chr.3: 15456190-15456190 on Build GRCh38
rs747882952 Chr.3: 15456067-15456067 on Build GRCh38
rs754480859 Chr.3: 15455819-15455819 on Build GRCh38
rs770378854 Chr.3: 15455851-15455851 on Build GRCh38
rs759169923 Chr.3: 15455978-15455978 on Build GRCh38
rs924638382 Chr.3: 15456223-15456223 on Build GRCh38
rs116828761 Chr.3: 15456013-15456013 on Build GRCh38
rs969812260 Chr.3: 15456265-15456265 on Build GRCh38
rs960203238 Chr.3: 15456192-15456192 on Build GRCh38
rs548322139 Chr.3: 15455909-15455909 on Build GRCh38
rs759779155 Chr.3: 15456122-15456122 on Build GRCh38
rs748636506 Chr.3: 15455846-15455846 on Build GRCh38
rs1011435175 Chr.3: 15456224-15456224 on Build GRCh38
rs780452818 Chr.3: 15456051-15456051 on Build GRCh38
rs368153249 Chr.3: 15455858-15455858 on Build GRCh38
rs886615963 Chr.3: 15455839-15455839 on Build GRCh38
rs562700516 Chr.3: 15455815-15455815 on Build GRCh38
rs762306354 Chr.3: 15455947-15455947 on Build GRCh38
rs774726658 Chr.3: 15455855-15455855 on Build GRCh38
rs771893784 Chr.3: 15456072-15456072 on Build GRCh38
rs73818509 Chr.3: 15455818-15455818 on Build GRCh38
rs1037807664 Chr.3: 15455884-15455884 on Build GRCh38
rs747829039 Chr.3: 15456060-15456060 on Build GRCh38
rs756508281 Chr.3: 15456007-15456007 on Build GRCh38
rs772370338 Chr.3: 15456047-15456047 on Build GRCh38
rs539100854 Chr.3: 15455933-15455933 on Build GRCh38
rs771047223 Chr.3: 15455921-15455921 on Build GRCh38
rs199930637 Chr.3: 15455900-15455900 on Build GRCh38
rs749863165 Chr.3: 15455870-15455870 on Build GRCh38
rs991543462 Chr.3: 15456198-15456198 on Build GRCh38
rs116503231 Chr.3: 15455939-15455939 on Build GRCh38
rs779104097 Chr.3: 15456015-15456015 on Build GRCh38
rs570213500 Chr.3: 15456088-15456088 on Build GRCh38

View Full Product Details