The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190274.1 | NP_001177203.1 | 16 - 17 | 2081 | 102 |
NM_025133.4 | NP_079409.3 | 16 - 17 | 1870 | 102 | |
XM_005264572.4 | XP_005264629.1 | 16 - 17 | 2075 | 102 | |
XM_005264573.4 | XP_005264630.1 | 16 - 17 | 2028 | 102 | |
XM_017005015.1 | XP_016860504.1 | 16 - 17 | 2028 | 102 | |
XM_017005016.1 | XP_016860505.1 | 16 - 17 | 1894 | 102 | |
XM_017005017.1 | XP_016860506.1 | 16 - 17 | 1894 | 102 | |
GenBank mRNA | AK022735.1 | - | 12 - 13 | 1383 | 102 |
AK025477.1 | - | 1 - 2 | 73 | 102 | |
AK097734.1 | - | 5 - 6 | 618 | 102 | |
AK223592.1 | - | 16 - 17 | 1965 | 102 | |
AK292877.1 | - | 16 - 17 | 1920 | 102 | |
AK304140.1 | - | 16 - 17 | 1785 | 102 | |
AK315108.1 | - | 16 - 17 | 1870 | 102 | |
AY827075.1 | - | 16 - 17 | 2008 | 102 | |
BC012728.2 | - | 2 - 3 | 154 | 102 | |
BC043258.1 | - | 16 - 17 | 1972 | 102 | |
BC130445.1 | - | 16 - 17 | 1806 | 102 | |
BC136480.1 | - | 16 - 17 | 1806 | 102 |