Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- ALPK1
- Entrez Gene ID
-
80216
- Gene Name
- alpha kinase 1
- Gene Aliases
-
8430410J10Rik, LAK
- Chromosome Location
-
Chr.4: 112286420 - 112442618 on Build GRCh38
View Full Product Details