The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271896.1 | NP_001258825.1 | 5 - 6 | 713 | 67 |
GenBank mRNA | AK022590.1 | - | 5 - 6 | 663 | 67 |