The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001201573.1 | NP_001188502.1 | 6 - 7 | 511 | 75 |
NM_001201574.1 | NP_001188503.1 | 3 - 4 | 414 | 75 | |
NM_025152.2 | NP_079428.2 | 8 - 9 | 747 | 75 | |
NR_120408.1 | 7 - 8 | 653 | 75 | ||
XM_011537181.2 | XP_011535483.1 | 5 - 6 | 682 | 75 | |
XM_011537182.2 | XP_011535484.1 | 9 - 10 | 721 | 75 | |
XM_011537184.2 | XP_011535486.1 | 3 - 4 | 597 | 75 | |
XM_017021667.1 | XP_016877156.1 | 7 - 8 | 668 | 75 | |
GenBank mRNA | AK022722.1 | - | 8 - 9 | 660 | 75 |
AK093789.1 | - | 3 - 4 | 414 | 75 | |
AK295326.1 | - | 8 - 9 | 708 | 75 | |
AK301447.1 | - | 6 - 7 | 511 | 75 | |
AK316445.1 | - | 8 - 9 | 737 | 75 | |
BC024919.1 | - | 8 - 9 | 673 | 75 | |
BX248028.1 | - | 7 - 8 | 640 | 75 | |
KJ534907.1 | - | 8 - 9 | 692 | 75 | |
KJ534909.1 | - | 8 - 9 | 692 | 75 |