Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 23922230-23922777 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPL15
Entrez Gene ID
6138
Gene Name
ribosomal protein L15
Gene Aliases
DBA12, EC45, L15, RPL10, RPLY10, RPYL10
UniGene
Hs.381219
Gene Chromosome Location
Chr.3: 23916804-23923696 on Build GRCh38
Transcripts
NM_001253384.1

SNP Detail

SNP ID Chromosome Location
rs1016473526 Chr.3: 23922664-23922664 on Build GRCh38
rs918257100 Chr.3: 23922481-23922481 on Build GRCh38
rs973791541 Chr.3: 23922480-23922480 on Build GRCh38
rs1012053350 Chr.3: 23922263-23922263 on Build GRCh38
rs1031058185 Chr.3: 23922606-23922606 on Build GRCh38
rs533638621 Chr.3: 23922680-23922680 on Build GRCh38
rs892575134 Chr.3: 23922751-23922751 on Build GRCh38
rs1041439447 Chr.3: 23922694-23922694 on Build GRCh38
rs1056323802 Chr.3: 23922512-23922512 on Build GRCh38
rs529230496 Chr.3: 23922679-23922679 on Build GRCh38
rs1024159277 Chr.3: 23922279-23922279 on Build GRCh38
rs951069585 Chr.3: 23922698-23922698 on Build GRCh38
rs903960945 Chr.3: 23922311-23922311 on Build GRCh38
rs560637023 Chr.3: 23922645-23922645 on Build GRCh38
rs111495111 Chr.3: 23922502-23922502 on Build GRCh38
rs551502791 Chr.3: 23922734-23922734 on Build GRCh38
rs1031046823 Chr.3: 23922349-23922349 on Build GRCh38
rs560950751 Chr.3: 23922491-23922491 on Build GRCh38
rs896318520 Chr.3: 23922262-23922262 on Build GRCh38
rs956653055 Chr.3: 23922353-23922353 on Build GRCh38
rs893939537 Chr.3: 23922738-23922738 on Build GRCh38
rs117795161 Chr.3: 23922407-23922407 on Build GRCh38
rs917562318 Chr.3: 23922713-23922713 on Build GRCh38
rs902974262 Chr.3: 23922460-23922460 on Build GRCh38
rs745305607 Chr.3: 23922636-23922636 on Build GRCh38
rs188139275 Chr.3: 23922437-23922437 on Build GRCh38
rs918661025 Chr.3: 23922633-23922633 on Build GRCh38
rs1032541358 Chr.3: 23922466-23922466 on Build GRCh38
rs965309301 Chr.3: 23922598-23922598 on Build GRCh38
rs368845468 Chr.3: 23922705-23922705 on Build GRCh38
rs976770344 Chr.3: 23922266-23922266 on Build GRCh38
rs896379731 Chr.3: 23922513-23922513 on Build GRCh38
rs903681111 Chr.3: 23922535-23922535 on Build GRCh38
rs918674026 Chr.3: 23922287-23922287 on Build GRCh38
rs747507565 Chr.3: 23922690-23922691 on Build GRCh38
rs1045345333 Chr.3: 23922718-23922718 on Build GRCh38
rs947948675 Chr.3: 23922521-23922521 on Build GRCh38
rs138987291 Chr.3: 23922395-23922395 on Build GRCh38
rs577865117 Chr.3: 23922383-23922383 on Build GRCh38
rs1000953830 Chr.3: 23922553-23922553 on Build GRCh38
rs918307675 Chr.3: 23922688-23922688 on Build GRCh38
rs529553715 Chr.3: 23922498-23922498 on Build GRCh38
rs1052537515 Chr.3: 23922750-23922750 on Build GRCh38
rs909775864 Chr.3: 23922362-23922362 on Build GRCh38
rs772056990 Chr.3: 23922743-23922743 on Build GRCh38
rs1000942407 Chr.3: 23922343-23922343 on Build GRCh38
rs1018949662 Chr.3: 23922586-23922586 on Build GRCh38
rs572194107 Chr.3: 23922436-23922436 on Build GRCh38
rs925148237 Chr.3: 23922733-23922733 on Build GRCh38
rs953688350 Chr.3: 23922404-23922404 on Build GRCh38
rs1030727549 Chr.3: 23922652-23922652 on Build GRCh38
rs948035288 Chr.3: 23922717-23922717 on Build GRCh38
rs756353886 Chr.3: 23922397-23922397 on Build GRCh38
rs1044934967 Chr.3: 23922534-23922534 on Build GRCh38
rs551152954 Chr.3: 23922655-23922655 on Build GRCh38
rs953680682 Chr.3: 23922653-23922653 on Build GRCh38
rs753465795 Chr.3: 23922337-23922337 on Build GRCh38
rs576411967 Chr.3: 23922431-23922431 on Build GRCh38
rs746986670 Chr.3: 23922654-23922654 on Build GRCh38
rs781173546 Chr.3: 23922282-23922282 on Build GRCh38
rs35693585 Chr.3: 23922671-23922671 on Build GRCh38
rs929807091 Chr.3: 23922333-23922333 on Build GRCh38

View Full Product Details