The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001272004.1 | NP_001258933.1 | 13 - 14 | 2678 | 67 |
NM_001272019.2 | NP_001258948.1 | 13 - 14 | 2284 | 67 | |
NM_001282391.1 | NP_001269320.1 | 13 - 14 | 2530 | 67 | |
NM_025209.3 | NP_079485.1 | 14 - 15 | 2747 | 67 | |
XM_006717510.1 | XP_006717573.1 | 14 - 15 | 2599 | 67 | |
GenBank mRNA | AF277374.1 | - | 14 - 15 | 2714 | 67 |
AF286905.1 | - | 13 - 14 | 2348 | 67 | |
AK024329.1 | - | 10 - 11 | 1793 | 67 | |
AK092304.1 | - | 13 - 14 | 2284 | 67 | |
AK299676.1 | - | 13 - 14 | 2471 | 67 | |
BC036529.1 | - | 13 - 14 | 2679 | 67 |