The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025212.3 | NP_079488.2 | 2 - 3 | 1514 | 92 |
XM_011532284.2 | XP_011530586.1 | 3 - 4 | 1656 | 92 | |
XM_017008652.1 | XP_016864141.1 | 2 - 3 | 1497 | 92 | |
GenBank mRNA | AF272159.1 | - | 1 - 2 | 564 | 92 |
AK127778.1 | - | 3 - 4 | 1311 | 92 | |
BC119751.1 | - | 1 - 2 | 564 | 92 | |
BC119752.1 | - | 1 - 2 | 564 | 92 |