The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025225.2 | NP_079501.2 | 8 - 9 | 1393 | 69 |
GenBank mRNA | AK025665.1 | - | 8 - 9 | 1393 | 69 |
AK123806.1 | - | 8 - 9 | 1317 | 69 | |
AK315166.1 | - | 8 - 9 | 1327 | 69 | |
AL138578.2 | - | 8 - 9 | 1393 | 69 | |
BC014449.1 | - | 3 - 4 | 322 | 69 | |
BC065195.1 | - | 8 - 9 | 1321 | 69 | |
CR456476.1 | - | 8 - 9 | 1221 | 69 |