The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025260.3 | NP_079536.2 | 3 - 4 | 539 | 81 |
NM_138272.2 | NP_612116.1 | 3 - 4 | 539 | 81 | |
NM_138277.2 | NP_612121.1 | 3 - 4 | 539 | 81 | |
XM_011514920.2 | XP_011513222.1 | 3 - 4 | 573 | 81 | |
XM_011514921.2 | XP_011513223.1 | 3 - 4 | 573 | 81 | |
GenBank mRNA | AJ292259.1 | - | 3 - 4 | 499 | 81 |
AJ292260.1 | - | 3 - 4 | 499 | 81 | |
AJ292264.1 | - | 3 - 4 | 499 | 81 | |
AJ292265.1 | - | 3 - 4 | 499 | 81 | |
AK092392.1 | - | 3 - 4 | 510 | 81 | |
BC113719.1 | - | 3 - 4 | 539 | 81 | |
BC113721.1 | - | 3 - 4 | 539 | 81 | |
BC143850.1 | - | 3 - 4 | 539 | 81 |