The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008695.1 | NP_001008695.1 | 3 - 4 | 265 | 101 |
NM_030573.2 | NP_085050.2 | 2 - 3 | 439 | 101 | |
GenBank mRNA | AK315457.1 | - | 2 - 3 | 424 | 101 |
BC004346.1 | - | 2 - 3 | 410 | 101 | |
BE264280.1 | - | 2 - 3 | 338 | 101 | |
BI223500.1 | - | 2 - 3 | 382 | 101 | |
BM790105.1 | - | 3 - 4 | 265 | 101 | |
CR456343.1 | - | 2 - 3 | 257 | 101 |