The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030775.2 | NP_110402.2 | 1 - 2 | 88 | 93 |
GenBank mRNA | AY009399.1 | - | 1 - 2 | 88 | 93 |