The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278919.1 | NP_001265848.1 | 5 - 6 | 1180 | 72 |
NM_001278920.1 | NP_001265849.1 | 5 - 6 | 1047 | 72 | |
NM_030779.3 | NP_110406.1 | 5 - 6 | 1180 | 72 | |
NM_173092.2 | NP_775115.1 | 5 - 6 | 1180 | 72 | |
XM_011525308.2 | XP_011523610.1 | 5 - 6 | 1180 | 72 | |
XM_011525309.2 | XP_011523611.1 | 5 - 6 | 1180 | 72 | |
XM_011525310.2 | XP_011523612.1 | 5 - 6 | 1180 | 72 | |
XM_011525311.1 | XP_011523613.1 | 5 - 6 | 1180 | 72 | |
XM_011525312.1 | XP_011523614.1 | 5 - 6 | 1180 | 72 | |
XM_011525313.1 | XP_011523615.1 | 5 - 6 | 1180 | 72 | |
XM_017025175.1 | XP_016880664.1 | 5 - 6 | 1180 | 72 | |
XM_017025176.1 | XP_016880665.1 | 5 - 6 | 1180 | 72 | |
XM_017025177.1 | XP_016880666.1 | 5 - 6 | 1180 | 72 | |
XM_017025178.1 | XP_016880667.1 | 5 - 6 | 1180 | 72 | |
XM_017025179.1 | XP_016880668.1 | 5 - 6 | 1180 | 72 | |
XM_017025180.1 | XP_016880669.1 | 5 - 6 | 1016 | 72 | |
GenBank mRNA | AF311913.1 | - | 5 - 6 | 1111 | 72 |
AK091877.1 | - | 5 - 6 | 1180 | 72 | |
AK296498.1 | - | 5 - 6 | 1153 | 72 | |
AK298363.1 | - | 5 - 6 | 1022 | 72 | |
BC006334.2 | - | 5 - 6 | 1158 | 72 |