The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030791.3 | NP_110418.1 | 1 - 2 | 837 | 75 |
GenBank mRNA | AF349315.1 | - | 1 - 2 | 759 | 75 |
AJ293294.1 | - | 1 - 2 | 718 | 75 | |
AK314188.1 | - | 1 - 2 | 783 | 75 | |
BC063839.1 | - | 1 - 2 | 780 | 75 | |
BC070060.1 | - | 1 - 2 | 797 | 75 |