The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135947.2 | NP_001129419.1 | 2 - 3 | 168 | 101 |
NM_001265582.1 | NP_001252511.1 | 2 - 3 | 168 | 101 | |
NM_030914.3 | NP_112176.1 | 2 - 3 | 168 | 101 | |
NR_049743.1 | 2 - 3 | 209 | 101 | ||
GenBank mRNA | AK001880.1 | - | 2 - 3 | 117 | 101 |
AK097029.1 | - | 2 - 3 | 114 | 101 | |
AK300877.1 | - | 2 - 3 | 115 | 101 | |
BC003581.1 | - | 2 - 3 | 126 | 101 | |
BI823604.1 | - | 2 - 3 | 168 | 101 | |
D83200.1 | - | 1 - 2 | 74 | 101 | |
DA691216.1 | - | 2 - 3 | 174 | 101 |