The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320273.1 | NP_001307202.1 | 3 - 4 | 635 | 71 |
NM_030965.2 | NP_112227.1 | 4 - 5 | 1045 | 71 | |
GenBank mRNA | AJ507292.1 | - | 4 - 5 | 789 | 71 |
AK056241.1 | - | 4 - 5 | 1012 | 71 | |
AK095758.1 | - | 5 - 6 | 931 | 71 | |
AK295019.1 | - | 5 - 6 | 888 | 71 | |
AK300902.1 | - | 3 - 4 | 560 | 71 | |
BC001201.1 | - | 4 - 5 | 952 | 71 |