The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190880.2 | NP_001177809.1 | 2 - 3 | 511 | 91 |
NM_031207.5 | NP_112484.3 | 2 - 3 | 511 | 91 | |
GenBank mRNA | AF284750.1 | - | 2 - 3 | 386 | 91 |
AF284751.1 | - | 2 - 3 | 386 | 91 | |
AY037165.1 | - | 2 - 3 | 485 | 91 | |
AY775560.1 | - | 2 - 3 | 486 | 91 | |
BC006140.1 | - | 2 - 3 | 197 | 91 | |
BC019041.2 | - | 2 - 3 | 313 | 91 | |
JF911821.1 | - | 1 - 2 | 96 | 91 |