The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031275.4 | NP_112565.1 | 2 - 3 | 192 | 98 |
GenBank mRNA | AF285600.1 | - | 1 - 2 | 98 | 98 |
BC029506.1 | - | 2 - 3 | 192 | 98 | |
BI561514.1 | - | 2 - 3 | 196 | 98 |