The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001003811.1 | NP_001003811.1 | 26 - 27 | 2347 | 87 |
NM_031276.2 | NP_112566.2 | 25 - 26 | 2225 | 87 | |
XM_011530994.1 | XP_011529296.1 | 25 - 26 | 2219 | 87 | |
XM_011530996.1 | XP_011529298.1 | 14 - 15 | 1365 | 87 | |
XM_017029649.1 | XP_016885138.1 | 25 - 26 | 2219 | 87 | |
XM_017029650.1 | XP_016885139.1 | 14 - 15 | 1411 | 87 | |
XM_017029651.1 | XP_016885140.1 | 14 - 15 | 1365 | 87 | |
XM_017029652.1 | XP_016885141.1 | 12 - 13 | 1046 | 87 | |
GenBank mRNA | AF285594.1 | - | 24 - 25 | 2130 | 87 |
AK057391.1 | - | 14 - 15 | 1366 | 87 | |
AK057523.1 | - | 26 - 27 | 2341 | 87 | |
AK292471.1 | - | 25 - 26 | 2219 | 87 | |
BC036016.1 | - | 25 - 26 | 2225 | 87 | |
BU929507.1 | - | 6 - 7 | 480 | 87 |