Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC70
Entrez Gene ID
83446
Gene Name
coiled-coil domain containing 70
Gene Aliases
-
Chromosome Location
Chr.13: 51861981 - 51866236 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031290.2 NP_112580.2 1 - 2 250 150
GenBank mRNA AK098719.1 - 1 - 2 250 150
AK225745.1 - 1 - 2 223 150
AL136887.1 - 1 - 2 221 150
BP371671.1 - 1 - 2 250 150

View Full Product Details