Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC25A31
Entrez Gene ID
83447
Gene Name
solute carrier family 25 member 31
Gene Aliases
AAC4, ANT 4, ANT4, SFEC35kDa
Chromosome Location
Chr.4: 127730375 - 127774299 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318467.1 NP_001305396.1 3 - 4 651 105
NM_031291.3 NP_112581.1 3 - 4 651 105
GenBank mRNA AJ863129.1 - 3 - 4 483 105
AL136857.1 - 3 - 4 610 105
AY550240.1 - 3 - 4 602 105
BC022032.2 - 3 - 4 602 105
BG773445.1 - 3 - 4 634 106
BM563946.1 - 3 - 4 643 105

View Full Product Details