Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CDCA3
Entrez Gene ID
83461
Gene Name
cell division cycle associated 3
Gene Aliases
GRCC8, TOME-1
Chromosome Location
Chr.12: 6844796 - 6852066 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001297603.1 NP_001284532.1 3 - 4 1154 82
NM_001297604.1 NP_001284533.1 3 - 4 1154 82
NM_031299.5 NP_112589.1 3 - 4 1154 82
NR_123727.1 3 - 4 1154 82
GenBank mRNA AK092246.1 - 3 - 4 357 82
AK291421.1 - 3 - 4 357 82
AK312843.1 - 3 - 4 374 82
BC002551.2 - 3 - 4 349 82
BX099267.1 - 2 - 3 322 82
CD106434.1 - 3 - 4 618 82
DA735127.1 - 3 - 4 347 82

View Full Product Details