The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270440.1 | NP_001257369.1 | 2 - 3 | 404 | 66 |
NM_031429.2 | NP_113617.1 | 2 - 3 | 404 | 66 | |
XM_005260089.1 | XP_005260146.1 | 2 - 3 | 404 | 66 | |
XM_017027343.1 | XP_016882832.1 | 2 - 3 | 404 | 66 | |
XM_017027344.1 | XP_016882833.1 | 2 - 3 | 404 | 66 | |
GenBank mRNA | AK093672.1 | - | 2 - 3 | 404 | 66 |
BC005063.1 | - | 2 - 3 | 392 | 66 | |
DB106146.1 | - | 2 - 3 | 404 | 66 |