The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031435.3 | NP_113623.1 | 1 - 2 | 687 | 81 |
GenBank mRNA | AK313452.1 | - | 1 - 2 | 249 | 81 |
AL136607.1 | - | 1 - 2 | 584 | 81 | |
BC008358.1 | - | 1 - 2 | 427 | 81 | |
CR533485.1 | - | 1 - 2 | 74 | 81 |