The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040177.2 | NP_001035267.1 | 2 - 3 | 358 | 65 |
NM_001271021.1 | NP_001257950.1 | 2 - 3 | 358 | 65 | |
NM_001271025.1 | NP_001257954.1 | 2 - 3 | 358 | 65 | |
NR_073125.1 | 2 - 3 | 358 | 65 | ||
NR_073126.1 | 2 - 3 | 272 | 65 | ||
NR_073127.1 | 2 - 3 | 358 | 65 | ||
XM_011519715.2 | XP_011518017.1 | 2 - 3 | 655 | 65 | |
XM_011519718.2 | XP_011518020.1 | 2 - 3 | 655 | 65 | |
XM_011519719.2 | XP_011518021.1 | 2 - 3 | 655 | 65 | |
XM_011519720.2 | XP_011518022.1 | 2 - 3 | 654 | 65 | |
XM_011519722.2 | XP_011518024.1 | 2 - 3 | 655 | 65 | |
XM_011519724.2 | XP_011518026.1 | 2 - 3 | 654 | 65 | |
XM_011519725.2 | XP_011518027.1 | 2 - 3 | 652 | 65 | |
XM_011519729.2 | XP_011518031.1 | 2 - 3 | 652 | 65 | |
XM_017016743.1 | XP_016872232.1 | 2 - 3 | 656 | 65 | |
XM_017016744.1 | XP_016872233.1 | 2 - 3 | 655 | 65 | |
XM_017016745.1 | XP_016872234.1 | 2 - 3 | 441 | 65 | |
GenBank mRNA | AB040820.1 | - | 2 - 3 | 239 | 65 |
AB040821.1 | - | 2 - 3 | 239 | 65 | |
AB040822.1 | - | 2 - 3 | 239 | 65 | |
AB055603.1 | - | 2 - 3 | 210 | 65 | |
AF263242.1 | - | 2 - 3 | 230 | 65 | |
AK074211.1 | - | 2 - 3 | 272 | 65 | |
BC002862.2 | - | 2 - 3 | 301 | 65 | |
DA006578.1 | - | 2 - 3 | 325 | 65 |