Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC3
Entrez Gene ID
83643
Gene Name
coiled-coil domain containing 3
Gene Aliases
-
Chromosome Location
Chr.10: 12896625 - 13099773 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031455.3 NP_113643.1 1 - 2 510 73
GenBank mRNA AL136562.1 - 1 - 2 426 73
BC051334.1 - 1 - 2 510 73

View Full Product Details