The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001287191.1 | NP_001274120.1 | 1 - 2 | 204 | 65 |
NM_001287192.1 | NP_001274121.1 | 1 - 2 | 204 | 65 | |
NM_031919.4 | NP_114125.1 | 1 - 2 | 204 | 65 | |
XM_005252257.3 | XP_005252314.1 | 1 - 2 | 199 | 65 | |
XM_005252260.3 | XP_005252317.1 | 1 - 2 | 189 | 65 | |
XM_017015183.1 | XP_016870672.1 | 1 - 2 | 199 | 65 | |
XM_017015187.1 | XP_016870676.1 | 1 - 2 | 199 | 65 | |
GenBank mRNA | AF088031.1 | - | 1 - 2 | 193 | 65 |
AF316830.1 | - | 1 - 2 | 67 | 65 | |
AK296433.1 | - | 1 - 2 | 115 | 65 | |
AK299491.1 | - | 1 - 2 | 115 | 65 | |
BP229804.1 | - | 1 - 2 | 204 | 65 | |
BP365251.1 | - | 1 - 2 | 194 | 65 |