The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031939.3 | NP_114145.2 | 2 - 3 | 291 | 85 |
XM_011526226.2 | XP_011524528.1 | 2 - 3 | 228 | 85 | |
XM_017026036.1 | XP_016881525.1 | 2 - 3 | 228 | 85 | |
GenBank mRNA | AB042647.1 | - | 2 - 3 | 155 | 85 |
AK054702.1 | - | 2 - 3 | 225 | 85 | |
AL705398.1 | - | 2 - 3 | 199 | 85 | |
BC029860.1 | - | 2 - 3 | 178 | 85 |