Product Details

Assay Reference Genome
Location

Chr.15:42775673 on build GRCh38
Cytoband
15q15.2
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
146057
Gene Symbol
TTBK2
Gene Name
tau tubulin kinase 2
Gene Aliases
SCA11, TTBK
Location
Chr.15:42738974-42920995 on build GRCh38
Assay Gene Location
Overlaps Intron 16 - Exon 17
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
TTBK2 NM_173500.3 NP_775771.3
XM_005254171.4 XP_005254228.1
XM_005254173.4 XP_005254230.1
XM_006720402.3 XP_006720465.1
XM_006720403.3 XP_006720466.1
XM_017021950.1 XP_016877439.1
AF525400.1 AAO14996.1
AK131372.1 BAD18523.1
BC071556.1 AAH71556.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv525461 Chr15:42770350 - 43189929 on Build GRCh38 Loss UBR1 TTBK2 CCNDBP1 TMEM62
nsv528725 Chr15:42463357 - 42812208 on Build GRCh38 Gain CDAN1 ZNF106 LRRC57 TTBK2 STARD9 SNAP23 HAUS2

View Full Product Details