Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM172A
Entrez Gene ID
83989
Gene Name
family with sequence similarity 172 member A
Gene Aliases
C5orf21
Chromosome Location
Chr.5: 93615130 - 94111699 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163417.1 NP_001156889.1 5 - 6 649 115
NM_032042.5 NP_114431.2 6 - 7 773 115
XM_005272103.3 XP_005272160.1 6 - 7 739 115
XM_005272108.3 XP_005272165.1 5 - 6 667 115
XM_005272109.4 XP_005272166.1 6 - 7 739 115
XM_005272111.4 XP_005272168.1 6 - 7 739 115
XM_006714717.3 XP_006714780.1 6 - 7 765 115
XM_006714718.3 XP_006714781.1 4 - 5 1379 115
XM_006714719.3 XP_006714782.1 4 - 5 528 115
XM_011543668.2 XP_011541970.1 6 - 7 739 115
XM_011543670.2 XP_011541972.1 6 - 7 739 115
XM_011543671.2 XP_011541973.1 6 - 7 739 115
XM_011543672.2 XP_011541974.1 6 - 7 669 115
XM_011543673.2 XP_011541975.1 5 - 6 568 115
XM_011543674.2 XP_011541976.1 6 - 7 739 115
XM_017009952.1 XP_016865441.1 6 - 7 1061 115
XM_017009953.1 XP_016865442.1 6 - 7 1059 115
XM_017009954.1 XP_016865443.1 6 - 7 739 115
XM_017009955.1 XP_016865444.1 5 - 6 1477 115
XM_017009956.1 XP_016865445.1 5 - 6 791 115
XM_017009957.1 XP_016865446.1 4 - 5 745 115
XM_017009958.1 XP_016865447.1 4 - 5 540 115
XM_017009959.1 XP_016865448.1 5 - 6 818 115
XM_017009962.1 XP_016865451.1 5 - 6 605 115
GenBank mRNA AK295127.1 - 5 - 6 672 115
AK296986.1 - 5 - 6 626 115
AK312930.1 - 6 - 7 678 115
AL136630.1 - 6 - 7 612 115
BC020584.2 - 5 - 6 575 115

View Full Product Details