Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
THRB
Entrez Gene ID
7068
Gene Name
thyroid hormone receptor beta
Gene Aliases
C-ERBA-2, C-ERBA-BETA, ERBA2, GRTH, NR1A2, PRTH, THR1, THRB1, THRB2
Chromosome Location
Chr.3: 24117153 - 24495281 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000461.4 NP_000452.2 6 - 7 820 79
NM_001128176.2 NP_001121648.1 7 - 8 931 79
NM_001128177.1 NP_001121649.1 7 - 8 957 79
NM_001252634.1 NP_001239563.1 8 - 9 1055 79
XM_005265421.4 XP_005265478.1 7 - 8 1028 79
XM_005265423.4 XP_005265480.1 6 - 7 863 79
XM_005265424.3 XP_005265481.1 6 - 7 798 79
XM_006713317.3 XP_006713380.1 7 - 8 1171 79
XM_006713318.3 XP_006713381.1 7 - 8 1123 79
XM_011534046.2 XP_011532348.1 7 - 8 3071 79
XM_011534047.2 XP_011532349.1 8 - 9 1308 79
XM_011534048.2 XP_011532350.1 7 - 8 887 79
XM_011534049.2 XP_011532351.1 6 - 7 743 79
XM_011534050.2 XP_011532352.1 7 - 8 1116 79
XM_011534051.2 XP_011532353.1 6 - 7 706 79
XM_011534052.2 XP_011532354.1 7 - 8 843 79
XM_011534053.2 XP_011532355.1 6 - 7 964 79
XM_017007108.1 XP_016862597.1 8 - 9 1241 79
XM_017007109.1 XP_016862598.1 9 - 10 1399 79
XM_017007110.1 XP_016862599.1 8 - 9 1206 79
XM_017007111.1 XP_016862600.1 7 - 8 852 79
XM_017007112.1 XP_016862601.1 6 - 7 822 79
XM_017007113.1 XP_016862602.1 5 - 6 961 79
XM_017007114.1 XP_016862603.1 5 - 6 926 79
XM_017007115.1 XP_016862604.1 5 - 6 800 79
GenBank mRNA AB307687.1 - 3 - 4 520 79
AK096628.1 - 8 - 9 1044 79
BC106929.1 - 5 - 6 645 79
BC106930.1 - 6 - 7 791 79
HQ692825.1 - 4 - 5 535 79
M26747.1 - 6 - 7 820 79
X04707.1 - 6 - 7 820 79

View Full Product Details