Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 46860840-46861342 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYL3
Entrez Gene ID
4634
Gene Name
myosin light chain 3
Gene Aliases
CMH8, MLC-lV/sb, MLC1SB, MLC1V, VLC1, VLCl
UniGene
Hs.517939
Gene Chromosome Location
Chr.3: 46857867-46863483 on Build GRCh38
Transcripts
NM_000258.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5728555 c.157G>A p.E53K Chr.3: 46860960-46860960 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs965440800 Chr.3: 46861308-46861308 on Build GRCh38
rs114333613 Chr.3: 46861132-46861132 on Build GRCh38
rs192329378 Chr.3: 46861001-46861001 on Build GRCh38
rs926402270 Chr.3: 46861057-46861057 on Build GRCh38
rs892221351 Chr.3: 46861225-46861225 on Build GRCh38
rs759706176 Chr.3: 46861034-46861034 on Build GRCh38
rs978037097 Chr.3: 46861110-46861110 on Build GRCh38
rs975702480 Chr.3: 46861078-46861078 on Build GRCh38
rs188566911 Chr.3: 46860871-46860871 on Build GRCh38
rs763012715 Chr.3: 46860862-46860862 on Build GRCh38
rs1009764276 Chr.3: 46861252-46861252 on Build GRCh38
rs1019357532 Chr.3: 46861269-46861269 on Build GRCh38
rs113513396 Chr.3: 46861165-46861165 on Build GRCh38
rs769273147 Chr.3: 46861018-46861018 on Build GRCh38
rs781740041 Chr.3: 46860921-46860921 on Build GRCh38
rs372306725 Chr.3: 46861031-46861031 on Build GRCh38
rs778515428 Chr.3: 46860977-46860977 on Build GRCh38
rs554596780 Chr.3: 46860886-46860886 on Build GRCh38
rs931498399 Chr.3: 46861094-46861094 on Build GRCh38
rs184579246 Chr.3: 46861149-46861149 on Build GRCh38
rs903860936 Chr.3: 46860941-46860941 on Build GRCh38
rs1042816943 Chr.3: 46860934-46860934 on Build GRCh38
rs1028593004 Chr.3: 46861318-46861318 on Build GRCh38
rs767888942 Chr.3: 46861036-46861036 on Build GRCh38
rs925163655 Chr.3: 46861115-46861115 on Build GRCh38
rs1004231349 Chr.3: 46860970-46860970 on Build GRCh38
rs899409586 Chr.3: 46861253-46861253 on Build GRCh38
rs542733581 Chr.3: 46860876-46860876 on Build GRCh38
rs905088251 Chr.3: 46861209-46861209 on Build GRCh38
rs997708651 Chr.3: 46860989-46860989 on Build GRCh38
rs77221239 Chr.3: 46861309-46861309 on Build GRCh38
rs995155437 Chr.3: 46861286-46861286 on Build GRCh38
rs752977796 Chr.3: 46860926-46860926 on Build GRCh38
rs56817407 Chr.3: 46861056-46861056 on Build GRCh38
rs567293216 Chr.3: 46861297-46861297 on Build GRCh38
rs776412048 Chr.3: 46861019-46861019 on Build GRCh38
rs1048565189 Chr.3: 46861107-46861107 on Build GRCh38
rs551103625 Chr.3: 46861088-46861088 on Build GRCh38
rs1057142162 Chr.3: 46861146-46861146 on Build GRCh38
rs931465740 Chr.3: 46860900-46860900 on Build GRCh38
rs895840157 Chr.3: 46861147-46861147 on Build GRCh38
rs777555567 Chr.3: 46860952-46860952 on Build GRCh38
rs562698157 Chr.3: 46861016-46861016 on Build GRCh38
rs921464842 Chr.3: 46860899-46860899 on Build GRCh38
rs886614220 Chr.3: 46861222-46861222 on Build GRCh38
rs992439903 Chr.3: 46861074-46861074 on Build GRCh38
rs549111363 Chr.3: 46861224-46861224 on Build GRCh38
rs569244804 Chr.3: 46861210-46861210 on Build GRCh38
rs1044987006 Chr.3: 46861208-46861208 on Build GRCh38
rs908644881 Chr.3: 46861117-46861117 on Build GRCh38
rs1039903239 Chr.3: 46860865-46860865 on Build GRCh38
rs749017586 Chr.3: 46860965-46860965 on Build GRCh38
rs376626859 Chr.3: 46861033-46861033 on Build GRCh38
rs374988162 Chr.3: 46860997-46860997 on Build GRCh38
rs1000639185 Chr.3: 46861026-46861026 on Build GRCh38
rs730880953 Chr.3: 46860981-46860981 on Build GRCh38
rs372045422 Chr.3: 46860951-46860951 on Build GRCh38
rs113179442 Chr.3: 46861064-46861064 on Build GRCh38
rs777261608 Chr.3: 46861039-46861039 on Build GRCh38
rs865996813 Chr.3: 46861168-46861168 on Build GRCh38

View Full Product Details