The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001987.4 | NP_001978.1 | 5 - 6 | 1278 | 61 |
XM_011520607.2 | XP_011518909.1 | 5 - 6 | 1528 | 61 | |
XM_011520608.2 | XP_011518910.1 | 5 - 6 | 1867 | 61 | |
XM_011520609.2 | XP_011518911.1 | 5 - 6 | 2142 | 61 | |
XM_011520611.2 | XP_011518913.1 | 4 - 5 | 1075 | 61 | |
XM_011520612.2 | XP_011518914.1 | 3 - 4 | 809 | 61 | |
XM_017018990.1 | XP_016874479.1 | 4 - 5 | 1396 | 61 | |
XM_017018991.1 | XP_016874480.1 | 9 - 10 | 2584 | 61 | |
GenBank mRNA | AK289441.1 | - | 5 - 6 | 1146 | 61 |
BC043399.1 | - | 5 - 6 | 1278 | 61 | |
KU178049.1 | - | 5 - 6 | 1004 | 61 | |
U11732.1 | - | 5 - 6 | 1028 | 61 |