Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TFAP4
Entrez Gene ID
7023
Gene Name
transcription factor AP-4 (activating enhancer binding protein 4)
Gene Aliases
AP-4, bHLHc41
Chromosome Location
Chr.16: 4257186 - 4273000 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003223.2 NP_003214.1 1 - 2 346 116
GenBank mRNA AK309301.1 - 1 - 2 413 116
BC010576.1 - 1 - 2 235 116
BU159415.1 - 1 - 2 299 116
CR541726.1 - 1 - 2 92 116
DA449763.1 - 1 - 2 413 116
S73885.1 - 1 - 2 350 116

View Full Product Details