Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TGIF1
Entrez Gene ID
7050
Gene Name
TGFB induced factor homeobox 1
Gene Aliases
HPE4, TGIF
Chromosome Location
Chr.18: 3411927 - 3458412 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278684.1 NP_001265613.1 2 - 3 608 139
NM_003244.3 NP_003235.1 1 - 2 341 139
NM_173208.2 NP_775300.1 2 - 3 611 139
XM_017025958.1 XP_016881447.1 2 - 3 672 139
GenBank mRNA AI813434.1 - 2 - 3 229 139
AK309471.1 - 1 - 2 336 139
AU310737.1 - 1 - 2 341 139
BC000814.1 - 1 - 2 107 139
BF446044.1 - 2 - 3 223 139
BQ437864.1 - 1 - 2 339 139
X89750.1 - 1 - 2 334 139

View Full Product Details