The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320793.1 | NP_001307722.1 | 4 - 5 | 611 | 75 |
NM_004379.4 | NP_004370.1 | 4 - 5 | 611 | 75 | |
NM_134442.4 | NP_604391.1 | 5 - 6 | 653 | 75 | |
NR_135473.1 | 5 - 6 | 653 | 75 | ||
XM_011510645.1 | XP_011508947.1 | 5 - 6 | 467 | 75 | |
XM_011510646.2 | XP_011508948.1 | 5 - 6 | 629 | 75 | |
XM_011510647.2 | XP_011508949.1 | 4 - 5 | 468 | 75 | |
XM_011510648.2 | XP_011508950.1 | 5 - 6 | 629 | 75 | |
XM_011510649.2 | XP_011508951.1 | 4 - 5 | 464 | 75 | |
XM_011510650.2 | XP_011508952.1 | 4 - 5 | 563 | 75 | |
XM_011510651.2 | XP_011508953.1 | 3 - 4 | 380 | 75 | |
XM_017003399.1 | XP_016858888.1 | 4 - 5 | 561 | 75 | |
XM_017003400.1 | XP_016858889.1 | 3 - 4 | 467 | 75 | |
XM_017003401.1 | XP_016858890.1 | 3 - 4 | 262 | 75 | |
GenBank mRNA | AK298740.1 | - | 4 - 5 | 433 | 75 |
AK302333.1 | - | 4 - 5 | 422 | 75 | |
AK312747.1 | - | 6 - 7 | 548 | 75 | |
AK315226.1 | - | 4 - 5 | 402 | 75 | |
AY347527.1 | - | 5 - 6 | 445 | 75 | |
BC010636.1 | - | 5 - 6 | 496 | 75 | |
BC095407.1 | - | 4 - 5 | 555 | 75 | |
BT019509.1 | - | 4 - 5 | 402 | 75 | |
CR407617.1 | - | 3 - 4 | 360 | 75 | |
M27691.1 | - | 4 - 5 | 476 | 75 | |
M34356.1 | - | 4 - 5 | 402 | 75 | |
X55545.1 | - | - | 545 | 75 | |
X60003.1 | - | 3 - 4 | 360 | 75 |