Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ELF3
Entrez Gene ID
1999
Gene Name
E74 like ETS transcription factor 3
Gene Aliases
EPR-1, ERT, ESE-1, ESX
Chromosome Location
Chr.1: 202010519 - 202017188 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114309.1 NP_001107781.1 7 - 8 1071 74
NM_004433.4 NP_004424.3 7 - 8 957 74
GenBank mRNA AF016295.1 - 7 - 8 918 74
AF017307.1 - 7 - 8 1006 74
AF517841.1 - 7 - 8 939 74
AK225446.1 - 7 - 8 894 74
AK310814.1 - 7 - 8 949 74
AK312273.1 - 7 - 8 933 74
BC003569.1 - 7 - 8 925 74
BX537368.1 - 6 - 7 3994 74
CR457106.1 - 6 - 7 804 74
U66894.1 - 7 - 8 899 74
U73843.1 - 7 - 8 923 74
U73844.1 - 8 - 9 854 74
U97156.1 - 7 - 8 928 74

View Full Product Details