Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TCF15
Entrez Gene ID
6939
Gene Name
transcription factor 15 (basic helix-loop-helix)
Gene Aliases
EC2, PARAXIS, bHLHa40
Chromosome Location
Chr.20: 603993 - 610266 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004609.3 NP_004600.2 1 - 2 558 102
GenBank mRNA AI218364.1 - 1 - 2 60 102
AW290965.1 - 1 - 2 59 101
BC067836.1 - 1 - 2 601 102
U08336.1 - 1 - 2 560 102

View Full Product Details