The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005334.2 | NP_005325.2 | 6 - 7 | 1436 | 72 |
XM_006724815.2 | XP_006724878.1 | 6 - 7 | 1901 | 72 | |
XM_006724816.2 | XP_006724879.1 | 6 - 7 | 1901 | 72 | |
XM_011531147.2 | XP_011529449.1 | 6 - 7 | 1901 | 72 | |
XM_011531148.2 | XP_011529450.1 | 6 - 7 | 1901 | 72 | |
XM_017029471.1 | XP_016884960.1 | 6 - 7 | 1901 | 72 | |
GenBank mRNA | BC019887.1 | - | 6 - 7 | 971 | 72 |
BC030560.1 | - | 6 - 7 | 1057 | 72 | |
BC063435.1 | - | 6 - 7 | 1447 | 72 | |
L20010.1 | - | 6 - 7 | 1252 | 72 |