The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204184.1 | NP_001191113.1 | 3 - 4 | 299 | 79 |
NM_001204185.1 | NP_001191114.1 | 3 - 4 | 299 | 79 | |
NM_001204186.1 | NP_001191115.1 | 3 - 4 | 299 | 79 | |
NM_001204187.1 | NP_001191116.1 | 3 - 4 | 299 | 79 | |
NM_001204188.1 | NP_001191117.1 | 3 - 4 | 299 | 79 | |
NM_005427.3 | NP_005418.1 | 3 - 4 | 299 | 79 | |
GenBank mRNA | BC117251.1 | - | 3 - 4 | 299 | 79 |
BC117253.1 | - | 3 - 4 | 299 | 79 | |
Y11416.1 | - | 3 - 4 | 299 | 79 |