The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005632.2 | NP_005623.1 | 2 - 3 | 228 | 111 |
XM_011522620.2 | XP_011520922.1 | 2 - 3 | 289 | 111 | |
XM_011522621.2 | XP_011520923.1 | 2 - 3 | 720 | 111 | |
XM_011522623.2 | XP_011520925.1 | 2 - 3 | 289 | 111 | |
XM_011522624.2 | XP_011520926.1 | 2 - 3 | 289 | 111 | |
XM_011522626.2 | XP_011520928.1 | 2 - 3 | 284 | 111 | |
XM_011522627.2 | XP_011520929.1 | 2 - 3 | 289 | 111 | |
XM_011522628.2 | XP_011520930.1 | 2 - 3 | 289 | 111 | |
XM_011522629.2 | XP_011520931.1 | 2 - 3 | 289 | 111 | |
XM_011522630.2 | XP_011520932.1 | 2 - 3 | 289 | 111 | |
XM_017023596.1 | XP_016879085.1 | 2 - 3 | 289 | 111 | |
GenBank mRNA | BC032648.1 | - | 2 - 3 | 142 | 111 |
BX341589.2 | - | 2 - 3 | 338 | 111 | |
U85647.1 | - | 2 - 3 | 228 | 111 |