The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037535.2 | NP_001032624.1 | 5 - 6 | 972 | 91 |
NM_001037536.2 | NP_001032625.1 | 5 - 6 | 969 | 91 | |
NM_001037540.2 | NP_001032629.1 | 7 - 8 | 1202 | 91 | |
NM_006746.5 | NP_006737.2 | 6 - 7 | 1121 | 91 | |
XM_005274578.2 | XP_005274635.1 | 7 - 8 | 1197 | 91 | |
XM_005274579.2 | XP_005274636.1 | 6 - 7 | 1110 | 91 | |
XM_006724508.3 | XP_006724571.1 | 6 - 7 | 4102 | 91 | |
XM_006724509.3 | XP_006724572.1 | 5 - 6 | 4018 | 91 | |
XM_011545564.2 | XP_011543866.1 | 6 - 7 | 4105 | 91 | |
XM_017029723.1 | XP_016885212.1 | 5 - 6 | 4021 | 91 | |
GenBank mRNA | AF160728.1 | - | 5 - 6 | 909 | 91 |
AK313993.1 | - | 5 - 6 | 930 | 91 | |
BC026159.2 | - | 6 - 7 | 1101 | 91 | |
BC105028.1 | - | 3 - 4 | 578 | 91 | |
BX647757.1 | - | 4 - 5 | 2886 | 91 |