The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130060.1 | NP_001123532.1 | 3 - 4 | 305 | 83 |
NM_001206682.1 | NP_001193611.1 | 3 - 4 | 305 | 83 | |
NM_001206683.1 | NP_001193612.1 | 3 - 4 | 214 | 83 | |
NM_006856.2 | NP_006847.1 | 3 - 4 | 305 | 83 | |
NR_073163.1 | 3 - 4 | 305 | 83 | ||
XM_005268587.3 | XP_005268644.1 | 3 - 4 | 257 | 83 | |
XM_017018722.1 | XP_016874211.1 | 3 - 4 | 1445 | 83 | |
GenBank mRNA | AK298853.1 | - | 3 - 4 | 251 | 83 |
AK313507.1 | - | 3 - 4 | 251 | 83 | |
BC136302.1 | - | 3 - 4 | 191 | 83 | |
BC140006.1 | - | 3 - 4 | 300 | 83 | |
DB047160.1 | - | 3 - 4 | 305 | 83 | |
X52943.1 | - | 3 - 4 | 250 | 83 | |
X57197.1 | - | 2 - 3 | 147 | 83 |